Stampa…

5-Chloro-2,1,3-benzothiadiazole 98%

Fornitore: Apollo Scientific

Synonyms: 5-Chlorobenzo-2,1,3-thiadiazole

OR909308-1G OR909308-5G
APOSOR909308-1GEA 53.9 EUR
APOSOR909308-1G APOSOR909308-5G
5-Chloro-2,1,3-benzothiadiazole 98%
5-Chloro-2,1,3-benzothiadiazole
Formula: C₆H₃ClN₂S
Peso molecolare: 170,62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00174269
CAS Number: 2207-32-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR