Stampa…

4-Methylbenzimidazole 95%

Fornitore: Apollo Scientific

Synonyms: 7-Methyl-1H-benzo[d]imidazole

OR909292-1G OR909292-250MG
APOSOR909292-1GEA 136 EUR
APOSOR909292-1G APOSOR909292-250MG
4-Methylbenzimidazole 95%
4-Methylbenzimidazole
Formula: C₈H₈N₂
Peso molecolare: 132,1600 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09842603
CAS Number: 4887-83-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR