Stampa…

1-Boc-piperidine 95%

Fornitore: Apollo Scientific

Synonyms: tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine

OR909204-100G OR909204-25G
APOSOR909204-100GEA 270 EUR
APOSOR909204-100G APOSOR909204-25G
1-Boc-piperidine 95%
1-Boc-piperidine
Formula: C₁₀H₁₉NO₂
Peso molecolare: 185,27 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02093935
CAS Number: 75844-69-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR