Stampa…

1-Chloro-3-methyl-5-nitrobenzene 97%

Fornitore: Apollo Scientific

Synonyms: 3-Chloro-5-nitrotoluene

OR908513-10G OR908513-25G
APOSOR908513-10GEA 80 EUR
APOSOR908513-10G APOSOR908513-25G
1-Chloro-3-methyl-5-nitrobenzene 97%
1-Chloro-3-methyl-5-nitrobenzene
Formula: C₇H₆ClNO₂
Peso molecolare: 171,58 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09955417
CAS Number: 16582-38-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR