Stampa…

4-Isopropoxyaniline 96%

Fornitore: Apollo Scientific

OR908467-100G OR908467-25G
APOSOR908467-100GEA 234 EUR
APOSOR908467-100G APOSOR908467-25G
4-Isopropoxyaniline 96%
4-Isopropoxyaniline
Formula: C₉H₁₃NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00059188
CAS Number: 7664-66-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR