2-(Propylthio)benzoic acid 98%
Fornitore: Apollo Scientific
Synonyms:
2-(Propylsulphanyl)benzoic acid
OR908020-10G
OR908020-1G
OR908020-5G
APOSOR908020-10GEA
502
EUR
APOSOR908020-10G
APOSOR908020-1G
APOSOR908020-5G
2-(Propylthio)benzoic acid 98%
2-(Propylthio)benzoic acid
Formula:
C₁₀H₁₂O₂S Peso molecolare: 196,27 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02729229 CAS Number: 21213-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...