Stampa…

3-Bromo-5-hydroxybenzonitrile 95%

Fornitore: Apollo Scientific

OR907983-1G OR907983-250MG
APOSOR907983-1GEA 115 EUR
APOSOR907983-1G APOSOR907983-250MG
3-Bromo-5-hydroxybenzonitrile 95%
3-Bromo-5-hydroxybenzonitrile
Formula: C₇H₄BrNO
Peso molecolare: 198,0200 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16659096
CAS Number: 770718-92-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR