Stampa…

4-Methyl-N-tert-butylbenzamide 98%

Fornitore: Apollo Scientific

Synonyms: N-tert-Butyl-4-methylbenzamide

OR907674-100G OR907674-10G OR907674-25G
APOSOR907674-100GEA 633 EUR
APOSOR907674-100G APOSOR907674-10G APOSOR907674-25G
4-Methyl-N-tert-butylbenzamide 98%
4-Methyl-N-tert-butylbenzamide
Formula: C₁₂H₁₇NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00463097
CAS Number: 42498-32-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR