Stampa…

2-Methylsulphanylpyrimidine-4-carbaldehyde 95%

Fornitore: Apollo Scientific

Synonyms: 2-(Methylthio)pyrimidine-4-carbaldehyde

OR907027-1G OR907027-250MG
APOSOR907027-1GEA 176 EUR
APOSOR907027-1G APOSOR907027-250MG
2-Methylsulphanylpyrimidine-4-carbaldehyde 95%
2-Methylsulphanylpyrimidine-4-carbaldehyde
Formula: C₆H₆N₂OS
Peso molecolare: 154,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02683211
CAS Number: 1074-68-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR