3'-Methoxypropiophenone 98%
Fornitore: Apollo Scientific
OR906997-100G
OR906997-25G
APOSOR906997-100GEA
114
EUR
APOSOR906997-100G
APOSOR906997-25G
3'-Methoxypropiophenone 98%
3'-Methoxypropiophenone
Formula:
C₁₀H₁₂O₂ Peso molecolare: 164,2011 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08458823 CAS Number: 37951-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...