Stampa…

4-Cyanobutanoic acid 97%

Fornitore: Apollo Scientific

OR906988-1G OR906988-250MG
APOSOR906988-1GEA 481 EUR
APOSOR906988-1G APOSOR906988-250MG
4-Cyanobutanoic acid 97%
4-Cyanobutanoic acid
Formula: C₅H₇NO₂
Peso molecolare: 113,12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00218781
CAS Number: 39201-33-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR