Stampa…

2-Methoxypyrimidine-4,6-diol 95%

Fornitore: Apollo Scientific

Synonyms: 4,6-Dihydroxy-2-methoxypyrimidine

OR906197-10G OR906197-1G OR906197-5G
APOSOR906197-10GEA 829 EUR
APOSOR906197-10G APOSOR906197-1G APOSOR906197-5G
2-Methoxypyrimidine-4,6-diol 95%
2-Methoxypyrimidine-4,6-diol
Formula: C₅H₆N₂O₃
Temperatura di stoccaggio: Ambiente
CAS Number: 1758-98-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR