Stampa…

α-Tropolone 98%

Fornitore: Apollo Scientific

Synonyms: Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone

OR906152-100G OR906152-25G
APOSOR906152-100GEA 611 EUR
APOSOR906152-100G APOSOR906152-25G
α-Tropolone 98%
α-Tropolone
Formula: C₇H₆O₂
Peso molecolare: 122,12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004158
CAS Number: 533-75-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR