Stampa…

N-Methylphthalimide 95%

Fornitore: Apollo Scientific

Synonyms: 2-Methylisoindoline-1,3-dione

OR906068-100G OR906068-25G OR906068-5G
APOSOR906068-100GEA 575 EUR
APOSOR906068-100G APOSOR906068-25G APOSOR906068-5G
N-Methylphthalimide 95%
N-Methylphthalimide
Formula: C₉H₇NO₂
Peso molecolare: 161,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023063
CAS Number: 550-44-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR