6-Cyano-1-tetralone 98%
Fornitore: Apollo Scientific
OR905894-1G
OR905894-250MG
OR905894-5G
APOSOR905894-1GEA
142
EUR
APOSOR905894-1G
APOSOR905894-250MG
APOSOR905894-5G
6-Cyano-1-tetralone 98%
6-Cyano-1-tetralone
Formula:
C₁₁H₉NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02179289 CAS Number: 90401-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...