Stampa…

5-Methyl-2-(piperazin-1-yl)thiazole 97%

Fornitore: Apollo Scientific

OR905841-1G OR905841-250MG
APOSOR905841-1GEA 807 EUR
APOSOR905841-1G APOSOR905841-250MG
5-Methyl-2-(piperazin-1-yl)thiazole 97%
5-Methyl-2-(piperazin-1-yl)thiazole
Formula: C8H13N3S
Temperatura di stoccaggio: Ambiente
CAS Number: 118113-05-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR