Stampa…

Methyl-2-methylthiophene-3-carboxylate 95%

Fornitore: Apollo Scientific

Synonyms: Methyl 2-methylthiophene-3-carboxylate

OR905711-1G OR905711-5G
APOSOR905711-1GEA 328 EUR
APOSOR905711-1G APOSOR905711-5G
Methyl-2-methylthiophene-3-carboxylate 95%
Methyl-2-methylthiophene-3-carboxylate
Formula: C₇H₈O₂S
Peso molecolare: 156,2000 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12026360
CAS Number: 53562-51-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR