2,4-Dichlorothiazole-5-carboxylic acid 95%
Fornitore: Apollo Scientific
OR905706-1G
OR905706-250MG
APOSOR905706-1GEA
224
EUR
APOSOR905706-1G
APOSOR905706-250MG
2,4-Dichlorothiazole-5-carboxylic acid 95%
2,4-Dichlorothiazole-5-carboxylic acid
Formula:
C₄HCl₂NO₂S Peso molecolare: 198,0300 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09909676 CAS Number: 62019-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...