Stampa…

2,5-Dichlorothiazole-4-carboxylic acid 95%

Fornitore: Apollo Scientific

OR905691-1G OR905691-250MG
APOSOR905691-1GEA 194 EUR
APOSOR905691-1G APOSOR905691-250MG
2,5-Dichlorothiazole-4-carboxylic acid 95%
2,5-Dichlorothiazole-4-carboxylic acid
Formula: C₄HCl₂NO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16615251
CAS Number: 127426-30-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR