Stampa…

Cycloheptyl isocyanide 95%

Fornitore: Apollo Scientific

OR905473-100MG OR905473-1G OR905473-250MG
APOSOR905473-100MGEA 190 EUR
APOSOR905473-100MG APOSOR905473-1G APOSOR905473-250MG
Cycloheptyl isocyanide 95%
Cycloheptyl isocyanide
Formula: C₈H₁₃N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02664622
CAS Number: 134420-07-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR