Stampa…

Chloromethyl isopropyl carbonate 97%

Fornitore: Apollo Scientific

Synonyms: Carbonic Acid Chloromethyl Isopropyl Ester

OR905009-100G OR905009-25G OR905009-500G
APOSOR905009-100GEA 0 EUR
APOSOR905009-100G APOSOR905009-25G APOSOR905009-500G
Chloromethyl isopropyl carbonate 97%
Chloromethyl isopropyl carbonate
Formula: C₅H₉ClO₃
Peso molecolare: 152,57 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07375443
CAS Number: 35180-01-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR