Stampa…

1-Piperidin-2-ylmethylazepane 96%

Fornitore: Apollo Scientific

OR904941-1G OR904941-5G
APOSOR904941-1GEA 290 EUR
APOSOR904941-1G APOSOR904941-5G
1-Piperidin-2-ylmethylazepane 96%
1-Piperidin-2-ylmethylazepane
Formula: C₁₂H₂₄N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05215076
CAS Number: 881040-50-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR