Stampa…

3,6-Dichloropyridazine-4-carbonitrile 98%

Fornitore: Apollo Scientific

OR904762-1G OR904762-250MG
APOSOR904762-1GEA 203 EUR
APOSOR904762-1G APOSOR904762-250MG
3,6-Dichloropyridazine-4-carbonitrile 98%
3,6-Dichloropyridazine-4-carbonitrile
Formula: C₅HCl₂N₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16621820
CAS Number: 35857-93-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR