2,5-Dimethyl-4-hydroxy-3(2H)-furanone 95%
Fornitore: Apollo Scientific
OR904751-100G
OR904751-500G
APOSOR904751-100GEA
132
EUR
APOSOR904751-100G
APOSOR904751-500G
2,5-Dimethyl-4-hydroxy-3(2H)-furanone 95%
2,5-Dimethyl-4-hydroxy-3(2H)-furanone
Formula:
C₆H₈O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00010706 CAS Number: 3658-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...