Stampa…

1-Pyridin-2-ylmethyl-1H-pyrrole-2-carbaldehyde 95%

Fornitore: Apollo Scientific

OR904571-1G OR904571-250MG
APOSOR904571-1GEA 684 EUR
APOSOR904571-1G APOSOR904571-250MG
1-Pyridin-2-ylmethyl-1H-pyrrole-2-carbaldehyde 95%
1-Pyridin-2-ylmethyl-1H-pyrrole-2-carbaldehyde
Formula: C₁₁H₁₀N₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02665150
CAS Number: 383135-91-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR