Stampa…

Acetohydrazide 96%

Fornitore: Apollo Scientific
Pericolo

Synonyms: Acetic hydrazide, Acethydrazide

OR904346-100G OR904346-500G
APOSOR904346-100GEA 84.4 EUR
APOSOR904346-100G APOSOR904346-500G
Acetohydrazide 96%
Acetohydrazide
Formula: C₂H₆N₂O
Peso molecolare: 74,08 g/mol
Punto di fusione: 58 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007610
CAS Number: 1068-57-1
UN: 2811
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR