Stampa…

6-Chloro-N,N-dimethylpyrimidin-4-amine 95%

Fornitore: Apollo Scientific

OR903838-1G OR903838-250MG
APOSOR903838-1GEA 87.3 EUR
APOSOR903838-1G APOSOR903838-250MG
6-Chloro-N,N-dimethylpyrimidin-4-amine 95%
6-Chloro-N,N-dimethylpyrimidin-4-amine
Formula: C₆H₈ClN₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09756954
CAS Number: 31058-83-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR