3-Chloro-5-methoxypyridazine
Fornitore: Apollo Scientific
OR903747-1G
OR903747-25G
OR903747-5G
APOSOR903747-1GEA
93.1
EUR
APOSOR903747-1G
APOSOR903747-25G
APOSOR903747-5G
3-Chloro-5-methoxypyridazine
3-Chloro-5-methoxypyridazine
Formula:
C5H5ClN2O Peso molecolare: 144,5600 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08236839 CAS Number: 123696-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...