Stampa…

6-Methoxybenzo[d]thiazol-2(3H)-one 95%

Fornitore: Apollo Scientific

Synonyms: 6-Methoxy-2(3H)-benzothiazolone

OR903746-1G OR903746-250MG
APOSOR903746-1GEA 364 EUR
APOSOR903746-1G APOSOR903746-250MG
6-Methoxybenzo[d]thiazol-2(3H)-one 95%
6-Methoxybenzo[d]thiazol-2(3H)-one
Formula: C₈H₇NO₂S
Peso molecolare: 181,2117 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09743851
CAS Number: 40925-65-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR