Stampa…

Methyl-3-quinolyl ether 95%

Fornitore: Apollo Scientific

Synonyms: 3-Methoxyquinoline

OR903566-1G OR903566-250MG OR903566-5G
APOSOR903566-1GEA 80 EUR
APOSOR903566-1G APOSOR903566-250MG APOSOR903566-5G
Methyl-3-quinolyl ether 95%
Methyl-3-quinolyl ether
Formula: C₁₀H₉NO
Peso molecolare: 159,1900 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16619425
CAS Number: 6931-17-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR