Stampa…

3,4-Dimethyl-1H-pyrazole-5-carboxylic acid 95%

Fornitore: Apollo Scientific

OR902641-100MG OR902641-1G OR902641-250MG
APOSOR902641-100MGEA 80 EUR
APOSOR902641-100MG APOSOR902641-1G APOSOR902641-250MG
3,4-Dimethyl-1H-pyrazole-5-carboxylic acid 95%
3,4-Dimethyl-1H-pyrazole-5-carboxylic acid
Formula: C6H8N2O2
Peso molecolare: 140,1400 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15143212
CAS Number: 89831-40-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR