Stampa…

Ethyl cyclohexanecarboxylate 95%

Fornitore: Apollo Scientific

OR902623-1G OR902623-25G OR902623-5G
APOSOR902623-1GEA 55.3 EUR
APOSOR902623-1G APOSOR902623-25G APOSOR902623-5G
Ethyl cyclohexanecarboxylate 95%
Ethyl cyclohexanecarboxylate
Formula: C₉H₁₆O₂
Peso molecolare: 156,23 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00041734
CAS Number: 3289-28-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR