5-Methyl-1,3-benzenediacetonitrile 95%
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₀N₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07782113 CAS Number: 120511-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...