1-(4-Thiazolyl)ethanone 98%
Fornitore: Apollo Scientific
OR902409-10G
OR902409-1G
OR902409-5G
OR902409-250MG
APOSOR902409-10GEA
242
EUR
APOSOR902409-10G
APOSOR902409-1G
APOSOR902409-5G
APOSOR902409-250MG
1-(4-Thiazolyl)ethanone 98%
1-(4-Thiazolyl)ethanone
Formula:
C₅H₅NOS Peso molecolare: 127,1643 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03550493 CAS Number: 38205-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...