3-Bromo-4,7-diazaindole 95%
Fornitore: Apollo Scientific
OR902271-100MG
OR902271-1G
OR902271-250MG
APOSOR902271-100MGEA
68.4
EUR
APOSOR902271-100MG
APOSOR902271-1G
APOSOR902271-250MG
3-Bromo-4,7-diazaindole 95%
3-Bromo-4,7-diazaindole
Formula:
C₆H₄BrN₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09834812 CAS Number: 56015-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...