Stampa…

Methyl-6-(hydroxymethyl)picolinate 95%

Fornitore: Apollo Scientific

Synonyms: Methyl 6-(hydroxymethyl)picolinate, Methyl 6-(hydroxymethyl)pyridine-2-carboxylate

OR902025-25G OR902025-5G
APOSOR902025-25GEA 355 EUR
APOSOR902025-25G APOSOR902025-5G
Methyl-6-(hydroxymethyl)picolinate 95%
Methyl-6-(hydroxymethyl)picolinate
Formula: C₈H₉NO₃
Peso molecolare: 167,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03840850
CAS Number: 39977-44-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR