Stampa…

Methyl-6-methoxypicolinate 98%

Fornitore: Apollo Scientific

OR901996-1G OR901996-5G
APOSOR901996-1GEA 0 EUR
APOSOR901996-1G APOSOR901996-5G
Methyl-6-methoxypicolinate 98%
Methyl-6-methoxypicolinate
Formula: C₈H₉NO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07779517
CAS Number: 26256-72-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR