Stampa…

Methyl-2-quinolyl ether 98%

Fornitore: Apollo Scientific

Synonyms: 2-Methoxyquinoline

OR901672-1G OR901672-5G
APOSOR901672-1GEA 120 EUR
APOSOR901672-1G APOSOR901672-5G
Methyl-2-quinolyl ether 98%
Methyl-2-quinolyl ether
Formula: C₁₀H₉NO
Peso molecolare: 159,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01070847
CAS Number: 6931-16-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR