Stampa…

Naphthalene-2-carboxamide 98%

Fornitore: Apollo Scientific

Synonyms: 2-Naphthylamide, 2-Naphthamide

OR900347-1G OR900347-25G OR900347-5G
APOSOR900347-1GEA 58.2 EUR
APOSOR900347-1G APOSOR900347-25G APOSOR900347-5G
Naphthalene-2-carboxamide 98%
Naphthalene-2-carboxamide
Formula: C₁₁H₉NO
Peso molecolare: 171,20 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016812
CAS Number: 2243-82-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR