5-(4-Bromophenoxy)-2-furoic acid
Fornitore: Apollo Scientific
OR8492-1G
OR8492-5G
OR8492-10G
APOSOR8492-1GEA
210
EUR
APOSOR8492-1G
APOSOR8492-5G
APOSOR8492-10G
5-(4-Bromophenoxy)-2-furoic acid
5-(4-Bromophenoxy)-2-furoic acid
Formula:
C₁₁H₇BrO₄ Punto di fusione: 173…175 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09607931 CAS Number: 73420-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...