Stampa…

Phenyl trans-styryl sulphone

Fornitore: Apollo Scientific
Attenzione

OR7693-1G OR7693-5G
APOSOR7693-1GEA 36.4 EUR
APOSOR7693-1G APOSOR7693-5G
Phenyl trans-styryl sulphone
Phenyl trans-styryl sulphone
Formula: C₁₄H₁₂O₂S
Punto di fusione: 74…76 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00159177
CAS Number: 16212-06-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR