7-{[(4-Methoxyphenyl)sulphonyl]amino}-1H-indole-2-carboxylic acid 95+%
Fornitore: Apollo Scientific
OR7670-1G
OR7670-5G
APOSOR7670-1GEA
209
EUR
APOSOR7670-1G
APOSOR7670-5G
7-{[(4-Methoxyphenyl)sulphonyl]amino}-1H-indole-2-carboxylic acid 95+%
7-{[(4-Methoxyphenyl)sulphonyl]amino}-1H-indole-2-carboxylic acid
Formula:
C₁₆H₁₄N₂O₅S Punto di fusione: 237…240 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01319295 CAS Number: 397245-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...