6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95+%
Fornitore: Apollo Scientific
OR7667-1G
OR7667-10G
OR7667-5G
APOSOR7667-1GEA
286
EUR
APOSOR7667-1G
APOSOR7667-10G
APOSOR7667-5G
6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95+%
6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one
Formula:
C₁₃H₁₂ClNO Peso molecolare: 233,7 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04037905 CAS Number: 849021-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...