2-Benzyloxy-5-chlorophenylboronic acid
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
(2-(Benzyloxy)-5-chlorophenyl)boronic acid
OR7153-1G
OR7153-5G
OR7153-10G
APOSOR7153-1GEA
81.5
EUR
APOSOR7153-1G
APOSOR7153-5G
APOSOR7153-10G
2-Benzyloxy-5-chlorophenylboronic acid
2-Benzyloxy-5-chlorophenylboronic acid
Formula:
C₁₃H₁₂BClO₃ Peso molecolare: 262,4966 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04039000 CAS Number: 612832-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...