1-(4-Bromophenyl)naphthalene
Fornitore: Apollo Scientific
|
Attenzione
|
OR70051-1G
OR70051-5G
APOSOR70051-1GEA
14.7
EUR
APOSOR70051-1G
APOSOR70051-5G
1-(4-Bromophenyl)naphthalene
1-(4-Bromophenyl)naphthalene
Formula:
C₁₆H₁₁Br Peso molecolare: 283,17 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07369916 CAS Number: 204530-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...