Stampa…

3-Bromo-7-nitro-1H-indazole

Fornitore: Apollo Scientific
Pericolo

Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

OR6862T-1G OR6862T-500MG OR6862T-5G
APOSOR6862T-1GEA 14.7 EUR
APOSOR6862T-1G APOSOR6862T-500MG APOSOR6862T-5G
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole

A more potent inhibitor of rat cerebellar nitric oxide synthase than 7-nitroindazole.Harmful to reproduction.

Formula: C₇H₄BrN₃O₂
Peso molecolare: 242,03 g/mol
Punto di fusione: 177…178 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR