Stampa…

Methyl (2E)-pent-2-enoate

Fornitore: Apollo Scientific
Pericolo

Synonyms: Methyl trans-2-pentenoate, (E)-Methyl pent-2-enoate

OR6667-25G OR6667-5G
APOSOR6667-25GEA 314 EUR
APOSOR6667-25G APOSOR6667-5G
Methyl (2E)-pent-2-enoate
Methyl (2E)-pent-2-enoate
Formula: C₆H₁₀O₂
Peso molecolare: 114,144 g/mol
Punto di ebollizione: 142…145 °C
Flash Pt: 16–26 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00137611
CAS Number: 15790-88-2
UN: 1993
ADR: 3,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR