Stampa…

LY-83583

Fornitore: Apollo Scientific
Attenzione

Synonyms: 6-Anilinoquinoline-5,8-quinone

OR6450T-25MG OR6450T-5MG
APOSOR6450T-25MGEA 350 EUR
APOSOR6450T-25MG APOSOR6450T-5MG
LY-83583
LY-83583

Inhibits nitric-oxide-induced activation of soluble guanylate cyclase(IC50=2uM)in a dose-dependent &reversible manner. It blocks acetylcholine inducedvasorelaxation.

Formula: C₁₅H₁₀N₂O₂
Peso molecolare: 250,25 g/mol
Punto di fusione: 184…185 °C
Temperatura di stoccaggio: Refrigerata
CAS Number: 91300-60-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR