Stampa…

3-Cyanophenyl isocyanate 97%

Fornitore: Apollo Scientific

Synonyms: 3-Cyanophenylisocyanate, 3-Isocyanatobenzonitrile

OR6433-1G OR6433-5G
APOSOR6433-1GEA 66.9 EUR
APOSOR6433-1G APOSOR6433-5G
3-Cyanophenyl isocyanate 97%
3-Cyanophenyl isocyanate

  • May form small amount of sediment.

Formula: C₈H₄N₂O
Peso molecolare: 144,13 g/mol
Punto di fusione: 51…54 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013856
CAS Number: 16413-26-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR