Stampa…

Acetohydroxamic acid

Fornitore: Apollo Scientific
Pericolo

Synonyms: N-Hydroxyacetamide

OR6375T-25G OR6375T-5G
APOSOR6375T-25GEA 48.1 EUR
APOSOR6375T-25G APOSOR6375T-5G
Acetohydroxamic acid
Acetohydroxamic acid

A urease inhibitor.

Formula: C₂H₅NO₂
Peso molecolare: 75.0666 g/mol
Punto di fusione: 86…90 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009994
CAS Number: 546-88-3
EINECS: 208-913-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR